siRNA Details
Gene Symbol: Ctsj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017121.1 | NP_058817.1 | 4 | 399 | ||
XM_008771439.1 | XP_008769661.1 | 3 | 314 | |||
GenBank mRNA | AF310623.2 | - | 4 | 399 | ||
BC097263.1 | - | 4 | 382 | 7460052 | ||
L14776.1 | - | 1 | 31 |
