siRNA Details
Gene Symbol: Ctsj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017121.1 | NP_058817.1 | 5 | 535 | ||
XM_008771439.1 | XP_008769661.1 | 4 | 450 | |||
GenBank mRNA | AF310623.2 | - | 5 | 535 | ||
BC097263.1 | - | 5 | 518 | 7460052 |
