siRNA Details
Gene Symbol: Neu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017130.2 | NP_058826.2 | 3 | 971 | ||
XM_006245361.2 | XP_006245423.1 | 3 | 911 | |||
XM_006245362.2 | XP_006245424.1 | 3 | 1575 | |||
XM_006245363.2 | XP_006245425.1 | 3 | 922 | |||
XM_006245364.1 | XP_006245426.1 | 3 | 877 | |||
XM_006245365.2 | XP_006245427.1 | 3 | 918 | |||
XM_008767209.1 | XP_008765431.1 | 4 | 1103 | |||
XM_008767210.1 | XP_008765432.1 | 4 | 1049 | |||
XM_008767211.1 | XP_008765433.1 | 3 | 1057 | |||
GenBank mRNA | BC091154.1 | - | 3 | 900 | 7323285 | |
D16300.1 | - | 3 | 971 | |||
FQ213221.1 | - | 3 | 934 | |||
FQ216178.1 | - | 3 | 846 |
