siRNA Details
Gene Symbol: Drd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017140.1 | NP_058836.1 | 1 | 280 | ||
XM_006248313.2 | XP_006248375.2 | 2 | 497 | |||
GenBank mRNA | S62729.1 | - | 1 | 207 | ||
X53944.1 | - | 1 | 280 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
