siRNA Details
Gene Symbol: Asgr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017189.2 | NP_058885.1 | 4 | 451 | ||
XM_006246676.2 | XP_006246738.1 | 4 | 529 | |||
GenBank mRNA | AF230645.1 | - | 4 | 451 | ||
BC088099.1 | - | 4 | 331 | 7322417 | ||
FQ218583.1 | - | 2 | 157 | |||
J02762.1 | - | 4 | 451 | |||
M16347.1 | - | 4 | 430 | |||
X07636.1 | - | 3 | 354 |
