siRNA Details
Gene Symbol: Celf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083586.2 | NP_001077055.2 | 12 | 1511 | ||
NM_017197.3 | NP_058893.2 | 12 | 1529 | |||
XM_008771861.1 | XP_008770083.1 | 12 | 1408 | |||
XM_008771862.1 | XP_008770084.1 | 11 | 1966 | |||
XM_008771863.1 | XP_008770085.1 | 11 | 1959 | |||
XM_008771864.1 | XP_008770086.1 | 11 | 1947 | |||
GenBank mRNA | AF090695.1 | - | 12 | 1345 | ||
AF169013.1 | - | 4 | 435 | |||
AJ010351.1 | - | 12 | 1454 | |||
AJ010386.1 | - | 12 | 1577 | |||
BC129096.1 | - | 12 | 1337 | 8362847 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001110231, NM_001110228, NM_001110230, NM_001110229, NM_001110232, and others.
Gene Ontology Categories:
