siRNA Details
Gene Symbol: Trpv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270797.1 | NP_001257726.1 | 2 | 554 | ||
NM_001270798.1 | NP_001257727.1 | 2 | 493 | |||
NM_017207.3 | NP_058903.2 | 2 | 577 | |||
GenBank mRNA | AB022332.1 | - | 2 | 509 | ||
AB029330.1 | - | 2 | 476 | |||
AF129113.1 | - | 2 | 496 | |||
BC089215.1 | - | 2 | 509 | 7302251 |
