siRNA Details
Gene Symbol: Trpv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270797.1 | NP_001257726.1 | 3 | 703 | ||
NM_001270798.1 | NP_001257727.1 | 3 | 642 | |||
NM_017207.3 | NP_058903.2 | 3 | 726 | |||
GenBank mRNA | AB022332.1 | - | 3 | 658 | ||
AB029330.1 | - | 3 | 625 | |||
AF129113.1 | - | 3 | 645 | |||
BC089215.1 | - | 3 | 658 | 7302251 |

More Information
Other Species Targeted

Gene Ontology Categories:
