siRNA Details
Gene Symbol: Slc1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017215.2 | NP_058911.2 | 1821 | |||
GenBank mRNA | AF297648.1 | - | 1733 | |||
AY069978.1 | - | 1012 | ||||
AY578981.1 | - | 5 | 535 | |||
JN132400.1 | - | 1896 | ||||
U15098.1 | - | 1770 | ||||
X67857.1 | - | 1808 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
