siRNA Details
Gene Symbol: Slc10a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017222.2 | NP_058918.1 | 6 | 1138 | ||
GenBank mRNA | U07183.2 | - | 6 | 1138 |

More Information
Other Species Targeted

Gene Ontology Categories:
