siRNA Details
Gene Symbol: Pex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017234.1 | NP_058930.1 | 3 | 745 | ||
XM_006232157.2 | XP_006232219.1 | 2 | 1834 | |||
XM_006232158.1 | XP_006232220.1 | 3 | 597 | |||
XM_006232159.1 | XP_006232221.1 | 3 | 853 | |||
XM_006232160.2 | XP_006232222.1 | 3 | 1934 | |||
XM_006232161.1 | XP_006232223.1 | 3 | 863 | |||
XM_008760865.1 | XP_008759087.1 | 3 | 614 | |||
XM_008760866.1 | XP_008759088.1 | 2 | 510 | |||
GenBank mRNA | BC063169.1 | - | 3 | 595 | 6886755 | |
D30616.1 | - | 3 | 712 | |||
D30617.1 | - | 3 | 745 | |||
X57988.1 | - | 2 | 521 |
