siRNA Details
Gene Symbol: Myh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017240.2 | NP_058936.1 | 37, 38 | 5806 | ||
XM_006251951.2 | XP_006252013.1 | 39, 40 | 5956 | |||
GenBank mRNA | FQ215585.1 | - | 6, 7 | 1070 | ||
FQ216900.1 | - | 5, 6 | 837 | |||
FQ216901.1 | - | 6, 7 | 946 | |||
FQ216969.1 | - | 3, 4 | 319 | |||
FQ217025.1 | - | 4, 5 | 607 | |||
FQ217049.1 | - | 3, 4 | 523 | |||
FQ217078.1 | - | 4, 5 | 610 | |||
FQ217204.1 | - | 1, 2 | 91 | |||
FQ217214.1 | - | 8, 9 | 1460 | |||
FQ217258.1 | - | 3, 4 | 439 | |||
FQ217267.1 | - | 4, 5 | 607 | |||
FQ217344.1 | - | 4, 5 | 610 | |||
FQ217392.1 | - | 6, 7 | 1058 | |||
FQ217405.1 | - | 4, 5 | 607 | |||
FQ217522.1 | - | 6, 7 | 943 | |||
FQ217648.1 | - | 1, 2 | 37 | |||
FQ217657.1 | - | 2, 3 | 184 | |||
FQ217776.1 | - | 1, 2 | 88 | |||
FQ217816.1 | - | 4, 5 | 610 | |||
FQ217848.1 | - | 4, 5 | 610 | |||
FQ217941.1 | - | 3, 4 | 364 | |||
FQ217965.1 | - | 4, 5 | 607 | |||
FQ218011.1 | - | 4, 5 | 544 | |||
FQ218024.1 | - | 3, 4 | 319 | |||
FQ223551.1 | - | 3, 4 | 439 | |||
FQ223587.1 | - | 3, 4 | 364 | |||
FQ223634.1 | - | 6, 7 | 943 | |||
FQ223665.1 | - | 4, 5 | 607 | |||
FQ223806.1 | - | 4, 5 | 607 | |||
FQ223821.1 | - | 4, 5 | 610 | |||
FQ223829.1 | - | 6, 7 | 943 | |||
FQ223892.1 | - | 4, 5 | 607 | |||
FQ223928.1 | - | 3, 4 | 290 | |||
FQ223936.1 | - | 4, 5 | 540 | |||
FQ223945.1 | - | 7, 8 | 1183 | |||
FQ224001.1 | - | 3, 4 | 367 | |||
FQ224034.1 | - | 2, 3 | 184 | |||
FQ224069.1 | - | 3, 4 | 319 | |||
FQ224090.1 | - | 3, 4 | 439 | |||
FQ224098.1 | - | 3, 4 | 439 | |||
FQ224119.1 | - | 4, 5 | 535 | |||
FQ224260.1 | - | 5, 6 | 822 | |||
FQ224368.1 | - | 3, 4 | 364 | |||
FQ224375.1 | - | 2, 3 | 231 | |||
FQ224416.1 | - | 3, 4 | 439 | |||
FQ224696.1 | - | 4, 5 | 607 | |||
FQ224765.1 | - | 4, 5 | 564 | |||
FQ224790.1 | - | 3, 4 | 268 | |||
J00752.1 | - | 7, 8 | 1229 | |||
M32698.1 | - | 2, 3 | 188 | |||
X15939.1 | - | 37, 38 | 5808 |
