siRNA Details
Gene Symbol: Prps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017243.2 | NP_058939.1 | 3 | 480 | ||
GenBank mRNA | BC078853.1 | - | 3 | 478 | 7190069 | |
M17258.1 | - | 3 | 456 | |||
M29392.1 | - | 3 | 456 | |||
X16554.1 | - | 3 | 477 |

More Information
Other Species Targeted

Gene Ontology Categories:
