siRNA Details
Gene Symbol: TKTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145933.1 | NP_001139405.1 | 13 | 2181 | ||
NM_001145934.1 | NP_001139406.1 | 12 | 2126 | |||
NM_012253.3 | NP_036385.3 | 13 | 2199 | |||
GenBank mRNA | AK292261.1 | - | 13 | 2075 | ||
AK301724.1 | - | 13 | 2000 | |||
AK302044.1 | - | 13 | 2057 | |||
DB524246.1 | - | 1 | 19 | |||
X91817.1 | - | 12 | 1965 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
