siRNA Details
Gene Symbol: Timm44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017267.1 | NP_058963.1 | 4 | 351 | ||
GenBank mRNA | AB006452.1 | - | 4 | 351 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Timm44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017267.1 | NP_058963.1 | 4 | 351 | ||
GenBank mRNA | AB006452.1 | - | 4 | 351 |