siRNA Details
Gene Symbol: Gpam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017274.1 | NP_058970.1 | 11, 12 | 1388 | ||
XM_006231625.2 | XP_006231687.1 | 11, 12 | 1779 | |||
XM_006231626.2 | XP_006231688.1 | 12, 13 | 1660 | |||
XM_008760534.1 | XP_008758756.1 | 11, 12 | 1330 | |||
GenBank mRNA | AF021348.1 | - | 11, 12 | 1388 | ||
U36771.2 | - | 10, 11 | 1164 |
