siRNA Details
Gene Symbol: Psma6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017283.3 | NP_058979.1 | 3 | 260 | ||
XM_006240098.2 | XP_006240160.1 | 3 | 218 | |||
GenBank mRNA | BC062232.1 | - | 3 | 261 | 6888689 | |
D10755.1 | - | 3 | 249 | |||
FQ212808.1 | - | 362 | ||||
FQ217234.1 | - | 3 | 236 | |||
FQ220193.1 | - | 3 | 287 |
