siRNA Details
Gene Symbol: Scn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271045.1 | NP_001257974.1 | 5 | 767 | ||
NM_001271046.2 | NP_001257975.1 | 5 | 634 | |||
NM_017288.2 | NP_058984.1 | 5 | 848 | |||
GenBank mRNA | BC094523.1 | - | 5 | 767 | 7316686 | |
FQ215379.1 | - | 3 | 425 | |||
FQ216374.1 | - | 5 | 634 | |||
M91808.1 | - | 5 | 847 |
