siRNA Details
Gene Symbol: S1pr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017301.2 | NP_058997.1 | 2 | 535 | ||
XM_008761459.1 | XP_008759681.1 | 2 | 587 | |||
GenBank mRNA | BC097938.1 | - | 2 | 535 | 7460011 | |
U10303.1 | - | 571 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
