siRNA Details
Gene Symbol: Sema6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017308.1 | NP_059004.1 | 1, 2 | 143 | ||
XM_006232858.2 | XP_006232920.1 | 4, 5 | 555 | |||
XM_006232861.2 | XP_006232923.1 | 2, 3 | 417 | |||
XM_008761286.1 | XP_008759508.1 | 2, 3 | 3821 | |||
XM_008761287.1 | XP_008759509.1 | 2, 3 | 3168 | |||
XM_008761288.1 | XP_008759510.1 | 2, 3 | 2094 | |||
XM_008761289.1 | XP_008759511.1 | 2, 3 | 3673 | |||
XM_008761290.1 | XP_008759512.1 | 2, 3 | 2920 | |||
XM_008761291.1 | XP_008759513.1 | 2, 3 | 3251 | |||
XM_008761292.1 | XP_008759514.1 | 2, 3 | 2350 | |||
XM_008761293.1 | XP_008759515.1 | 3, 4 | 507 | |||
XM_008761294.1 | XP_008759516.1 | 1, 2 | 3979 | |||
GenBank mRNA | AB000817.2 | - | 1, 2 | 159 | ||
AB014074.1 | - | 1, 2 | 159 | |||
AF363971.1 | - | 1, 2 | 143 |
