siRNA Details
Gene Symbol: Pde3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017337.1 | NP_059033.1 | 1 | 711 | ||
GenBank mRNA | U38179.1 | - | 1 | 711 |

More Information
Other Species Targeted

Gene Ontology Categories:
