siRNA Details
Gene Symbol: Ptprs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019140.2 | NP_062013.1 | 21 | 3947 | ||
GenBank mRNA | BC105753.1 | - | 4322 | 5599711 | ||
L12329.1 | - | 16 | 2861 | |||
L19180.1 | - | 2698 | ||||
L19181.1 | - | 15 | 2698 | |||
L19933.1 | - | 15 | 2775 |
