siRNA Details
Gene Symbol: RHO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000539.3 | NP_000530.1 | 1 | 428 | ||
GenBank mRNA | BC112104.1 | - | 1 | 420 | 8327572 | |
BC112106.1 | - | 1 | 420 | 8327574 | ||
BX537381.1 | - | 1 | 427 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
