siRNA Details
Gene Symbol: Syn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019159.1 | NP_062032.1 | 11 | 1580 | ||
GenBank mRNA | M27926.1 | - | 11 | 1580 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
