siRNA Details
Gene Symbol: Snca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019169.2 | NP_062042.1 | 4, 5 | 478 | ||
GenBank mRNA | AF007758.1 | - | 3, 4 | 341 | ||
AY550005.1 | - | 3, 4 | 341 | |||
AY550006.1 | - | 3, 4 | 341 | |||
BC087682.1 | - | 4, 5 | 478 | 7321273 | ||
FQ212580.1 | - | 4, 5 | 455 | |||
FQ227096.1 | - | 4, 5 | 469 | |||
S73007.1 | - | 4, 5 | 409 |
