siRNA Details
Gene Symbol: Snca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019169.2 | NP_062042.1 | 5 | 488 | ||
GenBank mRNA | AF007758.1 | - | 4 | 351 | ||
AY550005.1 | - | 4 | 351 | |||
AY550006.1 | - | 4 | 351 | |||
BC087682.1 | - | 5 | 488 | 7321273 | ||
FQ212580.1 | - | 5 | 465 | |||
FQ227096.1 | - | 5 | 479 | |||
S73007.1 | - | 5 | 419 |
