siRNA Details
Gene Symbol: RAPGEF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012294.3 | NP_036426.3 | 11, 12 | 1053 | ||
XM_005249914.2 | XP_005249971.1 | 11, 12 | 1272 | |||
XM_005249916.1 | XP_005249973.1 | 11, 12 | 1216 | |||
XM_011515652.2 | XP_011513954.1 | 3, 4 | 3026 | |||
XM_017012837.1 | XP_016868326.1 | 9, 10 | 957 | |||
GenBank mRNA | AK295859.1 | - | 1, 2 | 538 | ||
BC039203.1 | - | 1, 2 | 364 | 4811483 | ||
BP349215.1 | - | 2, 3 | 111 | |||
D87467.1 | - | 1, 2 | 345 | |||
KU178490.1 | - | 1, 2 | 290 | |||
KU178491.1 | - | 1, 2 | 290 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
