siRNA Details
Gene Symbol: Fdft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019238.2 | NP_062111.1 | 2 | 200 | ||
XM_008770746.1 | XP_008768968.1 | 1 | 2847 | |||
GenBank mRNA | BC081810.1 | - | 2 | 200 | 7188993 | |
FQ213323.1 | - | 2 | 183 | |||
FQ219372.1 | - | 2 | 171 | |||
M95591.1 | - | 2 | 218 |
