siRNA Details
Gene Symbol: Scn8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019266.2 | NP_062139.2 | 26 | 6482 | ||
GenBank mRNA | AF049239.1 | - | 26 | 6482 | ||
AF049240.1 | - | 26 | 6512 | |||
L39018.1 | - | 26 | 6308 | |||
M27223.1 | - | 1354 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011323, NM_001077499, AK083220, AK137395.
Gene Ontology Categories:
