siRNA Details
Gene Symbol: Kcnn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019313.1 | NP_062186.1 | 5, 6 | 1154 | ||
XM_006252933.2 | XP_006252995.1 | 5, 6 | 1187 | |||
GenBank mRNA | AF000973.1 | - | 5, 6 | 1154 | ||
DQ137426.1 | - | 1, 2 | 109 | |||
U69885.1 | - | 5, 6 | 920 |
