siRNA Details
Gene Symbol: Maf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019318.1 | NP_062191.1 | 1 | 1933 | ||
GenBank mRNA | U56242.1 | - | 1 | 1933 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001025577, S74567.
Gene Ontology Categories:
