siRNA Details
Gene Symbol: Cntn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019329.2 | NP_062202.1 | 2 | 211 | ||
XM_006236968.2 | XP_006237030.1 | 3 | 399 | |||
XM_008763197.1 | XP_008761419.1 | 3 | 399 | |||
GenBank mRNA | U11031.1 | - | 2 | 212 |
