siRNA Details
Gene Symbol: Rgs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019340.1 | NP_062213.1 | 6 | 697 | ||
XM_006238260.2 | XP_006238322.1 | 13 | 1280 | |||
XM_006238263.2 | XP_006238325.1 | 15 | 1455 | |||
XM_006238264.2 | XP_006238326.1 | 13 | 1282 | |||
XM_006238265.2 | XP_006238327.1 | 6 | 2897 | |||
XM_008763800.1 | XP_008762022.1 | 2 | 83 | |||
GenBank mRNA | AB055153.1 | - | 6 | 697 | ||
BC085710.1 | - | 6 | 686 |
