siRNA Details
Gene Symbol: Rgs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019340.1 | NP_062213.1 | 9, 10 | 1131 | ||
XM_006238260.2 | XP_006238322.1 | 16, 17 | 1714 | |||
XM_006238263.2 | XP_006238325.1 | 18, 19 | 1889 | |||
XM_006238264.2 | XP_006238326.1 | 16, 17 | 1716 | |||
XM_006238265.2 | XP_006238327.1 | 9, 10 | 3331 | |||
XM_008763800.1 | XP_008762022.1 | 5, 6 | 517 | |||
GenBank mRNA | AB055153.1 | - | 9, 10 | 1131 | ||
BC085710.1 | - | 9, 10 | 1120 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_134257, NM_001081650, NM_019492, AJ250999, BC131680, and others.
Gene Ontology Categories:
