siRNA Details
Gene Symbol: Haao | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020076.2 | NP_064461.1 | 10 | 1221 | ||
XM_006239684.2 | XP_006239746.1 | 9 | 2057 | |||
GenBank mRNA | BC085739.1 | - | 10 | 1175 | 7108551 | |
D44494.1 | - | 10 | 1222 | |||
FQ219702.1 | - | 10 | 1235 |
