siRNA Details
Gene Symbol: Adam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020077.1 | NP_064462.1 | 5 | 338 | ||
GenBank mRNA | X99794.1 | - | 5 | 338 |

More Information
Other Species Targeted

Gene Ontology Categories:
