siRNA Details
Gene Symbol: Dnase2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021664.1 | NP_067696.1 | 1 | 236 | ||
XM_006233464.2 | XP_006233526.1 | 2 | 313 | |||
XM_006233465.2 | XP_006233527.1 | 2 | 340 | |||
XM_006233466.2 | XP_006233528.1 | 2 | 343 | |||
XM_006233468.2 | XP_006233530.1 | 2 | 724 | |||
XM_008761521.1 | XP_008759743.1 | 2 | 446 | |||
GenBank mRNA | AF178974.1 | - | 1 | 236 |
