siRNA Details
Gene Symbol: Prkcz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022507.1 | NP_071952.1 | 14 | 1526 | ||
XM_006239535.2 | XP_006239597.1 | 11 | 4021 | |||
XM_006239536.2 | XP_006239598.1 | 11 | 3227 | |||
XM_008764337.1 | XP_008762559.1 | 14 | 1427 | |||
GenBank mRNA | J04532.1 | - | 14 | 1526 | ||
M18332.1 | - | 11 | 1079 | |||
U27191.1 | - | 11 | 1291 |
