siRNA Details
Gene Symbol: ICMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012405.3 | NP_036537.1 | 4 | 606 | ||
XM_011541140.2 | XP_011539442.1 | 3 | 306 | |||
GenBank mRNA | AF064084.1 | - | 4 | 576 | ||
AK092890.1 | - | 5 | 547 | |||
BC017037.1 | - | 5 | 701 | |||
BC028168.1 | - | 4 | 562 | 5245125 | ||
CR541671.1 | - | 4 | 533 | |||
CR541711.1 | - | 4 | 533 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
