siRNA Details
Gene Symbol: Slc30a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022853.2 | NP_074044.1 | 1 | 380 | ||
GenBank mRNA | U17133.1 | - | 1 | 381 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc30a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022853.2 | NP_074044.1 | 1 | 380 | ||
GenBank mRNA | U17133.1 | - | 1 | 381 |