siRNA Details
Gene Symbol: Faah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024132.3 | NP_077046.1 | 7 | 969 | ||
GenBank mRNA | BC091148.1 | - | 970 | 7370226 | ||
U72497.1 | - | 957 |
Gene Symbol: LOC100911581 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750001.3 | XP_003750049.1 | 7 | 1314 |
