siRNA Details
Gene Symbol: Crhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EU012436.1 | - | 2 | 104 | ||
EU012437.1 | - | 2 | 104 | |||
EU012438.1 | - | 2 | 104 | |||
L24096.1 | - | 2 | 182 | |||
L25438.1 | - | 2 | 104 | |||
U53499.1 | - | 1 | 44 |
