siRNA Details
Gene Symbol: Crhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EU012436.1 | - | 2, 3 | 118 | ||
EU012437.1 | - | 2, 3 | 118 | |||
EU012438.1 | - | 2, 3 | 118 | |||
L24096.1 | - | 2, 3 | 196 | |||
L25438.1 | - | 2, 3 | 118 |
