siRNA Details
Gene Symbol: Cpt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031559.2 | NP_113747.2 | 12 | 1429 | ||
XM_006230695.1 | XP_006230757.1 | 13 | 1539 | |||
GenBank mRNA | BC072522.1 | - | 12 | 1429 | 7105825 | |
L07736.1 | - | 12 | 1483 | |||
U88294.1 | - | 11 | 1381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
