siRNA Details
Gene Symbol: Rps7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031570.2 | NP_113758.2 | 4 | 247 | ||
GenBank mRNA | BC060557.1 | - | 4 | 251 | 6888098 | |
BC168722.1 | - | 3 | 231 | 6922220 | ||
FQ210052.1 | - | 4 | 247 | |||
FQ211139.1 | - | 4 | 247 | |||
FQ212121.1 | - | 4 | 247 | |||
FQ217452.1 | - | 4 | 247 | |||
FQ220886.1 | - | 4 | 295 | |||
FQ221065.1 | - | 4 | 247 | |||
FQ221374.1 | - | 4 | 247 | |||
FQ221761.1 | - | 4 | 247 | |||
FQ221878.1 | - | 4 | 247 | |||
FQ221932.1 | - | 4 | 250 | |||
FQ222169.1 | - | 4 | 247 | |||
FQ222484.1 | - | 4 | 247 | |||
FQ222534.1 | - | 4 | 245 | |||
FQ222918.1 | - | 4 | 247 | |||
FQ222996.1 | - | 4 | 247 | |||
FQ223010.1 | - | 4 | 247 | |||
FQ223103.1 | - | 4 | 247 | |||
FQ223196.1 | - | 4 | 246 | |||
FQ223290.1 | - | 4 | 247 | |||
FQ223392.1 | - | 4 | 247 | |||
FQ223426.1 | - | 4 | 247 | |||
FQ223491.1 | - | 4 | 247 | |||
FQ228541.1 | - | 4 | 263 | |||
FQ229000.1 | - | 4 | 245 | |||
X53377.1 | - | 4 | 247 | |||
X56846.1 | - | 3 | 231 |
Gene Symbol: LOC100362830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729576.4 | XP_002729622.2 | 1 | 247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
