siRNA Details
Gene Symbol: PTGER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028292.1 | 4 | 1363 | |||
NR_028293.1 | 3 | 1336 | ||||
GenBank mRNA | AY429108.1 | - | 3 | 1105 | ||
D86097.1 | - | 4 | 1363 | |||
D86098.1 | - | 3 | 1336 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
