siRNA Details
Gene Symbol: Vapa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031631.2 | NP_113819.3 | 4, 5 | 653 | ||
GenBank mRNA | AF086630.1 | - | 4, 5 | 401 | ||
BC061875.1 | - | 4, 5 | 653 | 5598274 | ||
FQ228815.1 | - | 4, 5 | 510 | |||
FQ234023.1 | - | 4, 5 | 410 |
