siRNA Details
Gene Symbol: Vapa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031631.2 | NP_113819.3 | 4 | 596 | ||
XM_008767384.1 | XP_008765606.1 | 4 | 1149 | |||
GenBank mRNA | AF086630.1 | - | 4 | 344 | ||
BC061875.1 | - | 4 | 596 | 5598274 | ||
FQ228815.1 | - | 4 | 453 | |||
FQ234023.1 | - | 4 | 353 |
