siRNA Details
Gene Symbol: Dlg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031639.1 | NP_113827.1 | 16, 17 | 2307 | ||
XM_006257080.2 | XP_006257142.1 | 16, 17 | 2634 | |||
XM_006257081.2 | XP_006257143.1 | 15, 16 | 2579 | |||
XM_006257082.2 | XP_006257144.1 | 15, 16 | 2542 | |||
XM_006257083.2 | XP_006257145.1 | 14, 15 | 2486 | |||
XM_006257084.2 | XP_006257146.1 | 9, 10 | 1034 | |||
XM_006257085.2 | XP_006257147.1 | 7, 8 | 1425 | |||
XM_008773267.1 | XP_008771489.1 | 15, 16 | 2594 | |||
XM_008773268.1 | XP_008771490.1 | 9, 10 | 943 | |||
GenBank mRNA | U50147.1 | - | 16, 17 | 2307 |
