siRNA Details
Gene Symbol: LOC100364062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751952.3 | XP_003752000.1 | 1 | 294 |
Gene Symbol: LOC100362738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772937.1 | XP_008771159.1 | 1 | 334 |
Gene Symbol: Pkm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053297.2 | NP_445749.1 | 4 | 370 | ||
XM_006243187.2 | XP_006243249.1 | 4 | 600 | |||
XM_006243188.2 | XP_006243250.1 | 4 | 600 | |||
XM_006243190.2 | XP_006243252.1 | 5 | 672 | |||
GenBank mRNA | AY724474.1 | - | 4 | 367 | ||
BC061541.1 | - | 4 | 377 | 6888187 | ||
FQ223721.1 | - | 4 | 378 | |||
FQ224316.1 | - | 1 | 28 | |||
X15800.1 | - | 4 | 372 |
